missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B4GALT4 Antibody (OTI8B6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71385F
This item is not returnable.
View return policy
Description
B4GALT4 Monoclonal specifically detects B4GALT4 in Human samples. It is validated for Immunohistochemistry.
Specifications
| B4GALT4 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| B4GALT4 | |
| Recombinant protein fragment corresponding to amino acids 40-322 of human B4GALT4 (NP_003769) produced in E.coli. | |
| 0.1 mL | |
| 8702.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI8B6 | |
| Immunohistochemistry | |
| B4Gal-T4, beta-1,4-galactosyltransferase 4, Beta-1,4-GalTase 4, beta4Gal-T4, beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 4, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 4, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 4, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 4, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction