missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B4GALT3 Antibody (OTI1G9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71386F
This item is not returnable.
View return policy
Description
B4GALT3 Monoclonal antibody specifically detects B4GALT3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| B4GALT3 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI1G9 | |
| Western Blot | |
| b4Gal-T3, beta-1,4-galactosyltransferase 3, Beta-1,4-GalTase 3, Beta4Gal-T3, beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 3, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 3, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 3, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 3 | |
| Human recombinant protein fragment corresponding to amino acids 100-393 of human B4GALT3(NP_003770) produced in E.coli. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 8703 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction