missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B4GALT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109440
This item is not returnable.
View return policy
Description
Immunogen sequence: PGVLMGGRYT PPDCTPAQTV AVIIPFRHRE HHLRYWLHYL HPILRRQRLR YGVYVINQHG EDTFNRAKLL NVGFLEALKE DAAYDCFIFS DVDLVPMDDR NLYRCGDQPR HFAIAMD.
B4GALT2 is one of type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. The enzyme synthesizes N-acetyllactosamine in glycolipids and glycoproteins. Its substrate specificity is affected by alpha-lactalbumin but it is not expressed in lactating mammary tissue.
Specifications
| B4GALT2 | |
| Polyclonal | |
| Unconjugated | |
| B4galt2 | |
| 4-Gal; B4galt2; B4Gal-T2; B4Gal-T3; Beta-1,4-galactosyltransferase 2; beta-1,4-galactosyltransferase II; beta-1,4-GalTase 2; beta-4-GalT2; beta4Gal-T2; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 2; Beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase; Ggtb2; Lactose synthase A protein; N-acetyllactosamine synthase; Nal synthase; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 2; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 2; UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 2; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 2; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 313536, 53418, 8704 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.66 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O60909, Q9Z2Y2 | |
| B4galt2 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction