missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B4GALNT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539056
This item is not returnable.
View return policy
Description
GM2 and GD2 gangliosides are sialic acid-containing glycosphingolipids. GalNAc-T is the enzyme involved in the biosynthesis of G(M2) and G(D2) glycosphingolipids. GalNAc-T catalyzes the transfer of GalNAc into G(M3) and G(D3) by a beta-1,4 linkage, resulting in the synthesis of G(M2) and G(D2), respectively. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Feb 2013].
Specifications
| B4GALNT1 | |
| Polyclonal | |
| Unconjugated | |
| B4GALNT1 | |
| GALGT, SIAT2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14421, 2583, 64828 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q00973, Q09200, Q10468 | |
| B4GALNT1 | |
| A synthetic peptide derived from the internal region of human B4GALNT1 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction