missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B4GALNT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109439
This item is not returnable.
View return policy
Description
Immunogen sequence: STRDAPGLRL PLAPWAPPQS PRRPELPDLA PEPRYAHIPV RIKEQVVGLL AWNNCSCESS GGGLPLPFQK QVRAIDLTKA FDPAELRAAS ATREQEFQAF LSRSQSPADQ LLIAPANSPL QYPLQGVEVQ PLRSILVPGL SLQAASGQEV YQVNLTASLG TWDVAGEVTG VTLTGEGQAD LTLVSPGLDQ LNRQLQLVTY SSRSYQTNTA DTGAR.
GM2 and GD2 gangliosides are sialic acid-containing glycosphingolipids. GalNAc-T is the enzyme involved in the biosynthesis of G(M2) and G(D2) glycosphingolipids. GalNAc-T catalyzes the transfer of GalNAc into G(M3) and G(D3) by a beta-1,4 linkage, resulting in the synthesis of G(M2) and G(D2), respectively. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| B4GALNT1 | |
| Polyclonal | |
| Unconjugated | |
| B4GALNT1 | |
| (N-acetylneuraminyl)-galactosylglucosylceramide; 4933429D13Rik; B4galnt1; Beta-1,4 N-acetylgalactosaminyltransferase 1; beta1,4GalNAC-T; beta-1,4-N-acetyl-galactosaminyl transferase 1; beta-1,4-N-acetyl-galactosaminyltransferase 1; beta-4N-acetylgalactosaminyltransferase; GALGT; Galgt1; GalNAcT; Gal-NAc-T; GalNAc-T; GD2 synthase, GM2 synthase; Ggm2; Ggm-2; GM2/GD2 synthase; SIAT2; SPG26; UDP-Gal:betaGlcNAc beta-1,4-N-acetylgalactosaminyltransferase transferase 1; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide N-acetylgalactosaminyltransferase (GalNAc-T); UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide-beta-1, 4-N-acetylgalactosaminyltransferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2583 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.15 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q00973 | |
| B4GALNT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 26-240 of human B4GALNT1 (NP_001263398.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction