missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GNT6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21291-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| B3GNT6 | |
| Polyclonal | |
| Unconjugated | |
| B3GNT6 | |
| B3Gn T6, B3GNT6, Beta 1,3 Gn T6, Beta3Gn T6, BGnT 6, Core 3 synthase, IMAGE:4907098 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192134, 272411 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6ZMB0 | |
| B3gnt6 | |
| B3GNT6 Fusion Protein Ag15825 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction