missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B3GNT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558936
This item is not returnable.
View return policy
Description
Immunogen sequence: QQWFLQAPRS PREERSPQEE TPEGPTDA Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 43%.
The protein encoded by this gene is a beta-1,3-N-acetylglucosaminyltransferase that adds an N-acetylglucosamine moiety to N-acetylgalactosamine-modified serine or threonine. The encoded enzyme is responsible for creating the core 3 structure of O-glycans, which are important components of mucin-type glycoproteins.
Specifications
| B3GNT6 | |
| Polyclonal | |
| Unconjugated | |
| B3GNT6 | |
| acetylgalactosaminyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; B3GNT6; B3Gn-T6; BC039789; beta-1,3-Gn-T6; beta-1,3-N-acetylglucosaminyltransferase 6; beta-1,3-N-acetylglucosaminyltransferase protein; beta3Gn-T6; BGnT-6; core 3 synthase; Core3; Core-3; RGD1310926; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 (core 3 synthase) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192134 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZMB0 | |
| B3GNT6 | |
| Recombinant protein corresponding to Human B3GNT6. Recombinant protein control fragment (Product #RP-104289). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction