missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B3GNT4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563102
This item is not returnable.
View return policy
Description
Immunogen sequence: TAHQPFWAPP TPRHSRCPPN HTVSSASLSL PSRHRLFLTY RHCRNFSILL EPSGCSKDTF LLLAIK Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%.
This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase protein family. The encoded enzyme is involved in the biosynthesis of poly-N-acetyllactosamine chains and prefers lacto-N-neotetraose as a substrate. It is a type II transmembrane protein.
Specifications
| B3GNT4 | |
| Polyclonal | |
| Unconjugated | |
| B3GNT4 | |
| 1010001G17Rik; B3GNT4; B3GN-T4; beta-1,3-Gn-T4; beta-1,3-N-acetylglucosaminyltransferase 4; beta-1,3-N-acetylglucosaminyltransferase bGn-T4; beta3Gn-T4; BGnT-4; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 4; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4; UNQ1898/PRO4344 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79369 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C0J1 | |
| B3GNT4 | |
| Recombinant protein corresponding to Human B3GNT4. Recombinant protein control fragment (Product #RP-101687). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction