missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GNT2, Mouse anti-Human, Clone: 1A8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010678-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: NLPDRFKDFLLYLRCRNYSLLIDQPDKCAKKPFLLLAIKSLTPHFARRQAIRESWGQESNAGNQTVVRVFLLGQTPPEDNHPDLSDMLKFESEKHQDILMSpecifications
| B3GNT2 | |
| Monoclonal | |
| Unconjugated | |
| UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2 | |
| B3GN-T1/B3GN-T2/B3GNT/B3GNT-2/B3GNT1/BETA3GNT | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1A8 | |
| In 1x PBS, pH 7.4 | |
| NM_006577 | |
| B3GNT2 | |
| B3GNT2 (NP_006568, 111 a.a. ∼ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10678 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction