Learn More
B3GAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA597275
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: VHKDEGSDPR RETPPGADPR EYCTSDRDIV EVVRTEYVYT RPPPWSDTLP TIHVVTPTYS RPVQKAELTR MANTLLHVPN LHWLVVEDAP RRTPLTARLL RDTGLNYTHL HVETPRNYKL RGDARDPRIP RGTMQRNLAL RWLRETFPRN SSQPGVVYFA DDDNTYSLEL FEEMRSTRRV SVWPVAFVGG LRYEAPRVNG AGKVVGWKTV FDPHRPFAID MAGFAVNLRL ILQRSQAYFK LRGVKGGYQE SSLLRELVTL NDLEPKAANC TKILVWHTRT EKPVLVNEGK KGFTDPSVEI; Positive Samples: SW480, HT-1080, BT-474; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein
The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized.Specifications
CD57 | |
Polyclonal | |
Unconjugated | |
B3GAT1 | |
B3GAT1 | |
Recombit fusion protein containing a sequence corresponding to amino acids 35-334 of human B3GAT1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
2.03 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9P2W7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27087 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |