missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA586578
Additional Details : Weight : 0.01000kg
Description
The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized.Specifications
CD57 | |
Polyclonal | |
Unconjugated | |
B3GAT1 | |
Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1; Beta-1,3-glucuronyltransferase 1; Glucuronosyltransferase P; GlcAT-P; UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase; GlcUAT-P; B3GAT1; GLCATP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117108, 27087, 76898 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.2 | |
O35789, Q9P2W7 | |
B3gat1 | |
Synthetic peptide, corresponding to amino acids 3-50 of Human CD57. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |