missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GAT1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00027087-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MPKRRDILAIVLIVLPWTLLITVWHQSTLAPLLAVHKDEGSDPRRETPPGADPREYCTSDRDIVEVVRTEYVYTRPPPWSDTLPTIHVVTPTYSRPVQKAELTRMANTLLHVPNLHWLVVEDAPRRTPLTARLLRDTGLNYTHLHVETPRNYKLRGDARDPRIPRGTMQRNLALRWLRETFPRNSSQPGVVYFADDDNTYSLELFEEMRSTRRVSVWPVAFVGGLRYEAPRVNGAGKVVGWKTVFDPHRPFAIDMAGFAVNLRLILQRSQAYFKLRGVKGGYQESSLLRELVTLNDLEPKAANCTKILVWHTRTEKPVLVNEGKKGFTDPSVEISpecifications
| B3GAT1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| DQ895278.2 | |
| B3GAT1 | |
| B3GAT1 (ABM86204.1, 1 a.a. ∼ 334 a.a) full-length human protein. | |
| RUO | |
| 27087 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P) | |
| CD57/GLCATP/GlcAT-P/GlcUAT-P/HNK-1/HNK1/LEU7/NK-1 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction