Learn More
B3GAT1 Mouse anti-Human, Clone: HNK-1, Abnova™
Mouse monoclonal antibody raised against native human B3GAT1.
Brand: Abnova MAB13279.100ug
Description
The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized. [provided by RefSeq]
Specifications
| B3GAT1 | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| CD57/GLCATP/GlcAT-P/GlcUAT-P/HNK-1/HNK1/LEU7/NK-1 | |
| Mouse | |
| PEG precipitation | |
| RUO | |
| 27087 | |
| Store at 4°C. | |
| IgM κ |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| HNK-1 | |
| Mouse monoclonal antibody raised against native human B3GAT1. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| B3GAT1 | |
| Human peripheral blood mononuclear cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.