missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B3GALTL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101457
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total B3GALTL.
The protein encoded by this gene is a beta-1, 3-glucosyltransferase that transfers glucose to O-linked fucosylglycans on thrombospondin type-1 repeats (TSRs) of several proteins. The encoded protein is a type II membrane protein. Defects in this gene are a cause of Peters-plus syndrome (PPS).
Specifications
| B3GALTL | |
| Polyclonal | |
| Unconjugated | |
| B3GLCT | |
| B3galtl; B3GLCT; B3Glc-T; B3GTL; beta 1,3-galactosyltransferase-like; beta 3-glucosyltransferase; beta 3-glycosyltransferase-like; beta-1,3-glucosyltransferase; Beta3Glc-T; beta-3-glucosyltransferase; beta-3-glycosyltransferase-like; Gal-T; Gm1057; UDP-GAL:beta-GlcNAc beta-1,3-galactosyltransferase-like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 145173, 381694 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6Y288, Q8BHT6 | |
| B3GLCT | |
| A synthesized peptide derived from human B3GLCT(Accession Q6Y288), corresponding to amino acid residues S461-L498. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction