missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GALT6, Mouse anti-Human, Clone: 3E5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00126792-M06.100ug
This item is not returnable.
View return policy
Description
Sequence: RLSRDYLRAWHSEDVSLGAWLAPVDVQREHDPRFDTEYRSRGCSNQYLVTHKQSLEDMLEKHATLAREGRLCKREVQLRLSYVYDWSAPPSQCCQRREGIPSpecifications
| B3GALT6 | |
| Monoclonal | |
| Unconjugated | |
| UDP-Gal:betaGal beta 1,3-galactosyltransferase polypeptide 6 | |
| beta3GalT6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3E5 | |
| In 1x PBS, pH 7.4 | |
| NM_080605 | |
| B3GALT6 | |
| B3GALT6 (NP_542172, 229 a.a. ∼ 329 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 126792 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction