missing translation for 'onlineSavingsMsg'
Learn More

B3GALT6 Antibody, Novus Biologicals™

Product Code. 18429468 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429468 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429468 Supplier Novus Biologicals Supplier No. H00126792B01P50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

B3GALT6 Polyclonal antibody specifically detects B3GALT6 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GALT6
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAI60034.1
Gene Alias beta-1,3-galactosyltransferase 6, Beta-1,3-GalTase 6, Beta3GalT6, Beta3Gal-T6, EC 2.4.1.134, Galactosyltransferase II, Galactosylxylosylprotein 3-beta-galactosyltransferase, UDP-Gal:betaGal beta 1,3-galactosyltransferase polypeptide 6GAG GalTII, UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 6
Host Species Mouse
Immunogen B3GALT6 (AAI60034.1, 1 a.a. - 329 a.a.) full-length human protein. MKLLRRAWRRRAALGLGTLALCGAALLYLARCAAEPGDPRAMSGRSPPPPAPARAAAFLAVLVASAPRAAERRSVIRSTWLARRGAPGDVWARFAVGTAGLGAEERRALEREQARHGDLLLLPALRDAYENLTAKVLAMLAWLDEHVAFEFVLKADDDSFARLDALLAELRAREPARRRRLYWGFFSGRGRVKPGGRWREAAWQLCDYYLPYALGGGYVLSADLVHYLRLSRDYLRAWHSEDVSLGAWLAPVDVQREHDPRFDTEYRSRGCSNQYLVTHKQSLEDMLEKHATLAREGRLCKREVQLRLSYVYDWSAPPSQCCQRREGIP
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126792
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.