missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B3GALT5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563006
This item is not returnable.
View return policy
Description
Immunogen sequence: YSLNPFKEQS FVYKKDGNFL KLPDTDCRQT PPFLVLLVTS SHKQLAERMA IRQTW Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 68%.
B3GALT5 catalyzes the transfer of Gal to GlcNAc-based acceptors with a preference for the core3 O-linked glycan GlcNAc(beta1,3)GalNAc structure. Can use glycolipid LC3Cer as an efficient acceptor. (Uniprot).
Specifications
| B3GALT5 | |
| Polyclonal | |
| Unconjugated | |
| B3GALT5 | |
| 1190002B21Rik; AU045265; B3GALT5; b3Gal-T5; b3Galt-V; B3GalTx; B3gt5; B3T5; Beta-1,3-galactosyltransferase 5; beta-1,3-GalTase 5; beta-3-galactosyltransferase 5; beta3GalT5; beta3Gal-T5; beta-3-Gx-T5; GLCT5; homolog of C. elegans Bt toxin resistance gene bre-5; SSEA-3 synthase; Stage-specific embryonic antigen 3 synthase; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 5; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 5; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 5; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10317 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2C3 | |
| B3GALT5 | |
| Recombinant protein corresponding to Human B3GALT5. Recombinant protein control fragment (Product #RP-101480). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction