missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B3GALT4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560194
This item is not returnable.
View return policy
Description
Immunogen sequence: PFPPYASGTG YVLSASAVQL ILKVASRAPL LPLEDVFVGV SARRGGLAPT QCVKLAGATH YPLDRCCYGK FLLTSHRLDP WKMQEAWKLV GGSDGERTAP FCSWFQGVLG ILRCRA Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
This gene is a member of the beta-1, 3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesized by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). This gene is oriented telomere to centromere in close proximity to the ribosomal protein S18 gene. The functionality of the encoded protein is limited to ganglioseries glycolipid biosynthesis.
Specifications
| B3GALT4 | |
| Polyclonal | |
| Unconjugated | |
| B3galt4 | |
| B3galt4; b3Gal-T4; Beta-1,3-galactosyltransferase 4; beta-1,3-GalTase 4; beta-3-galactosyltransferase 4; beta3GalT4; beta3Gal-T4; GALT2; Gal-T2; GALT4; ganglioside galactosyltransferase; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 4; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 4; UDP-Gal:betaGlcNAc beta 13-galactosyltransferase polypeptide 4; UDP-galactose:beta-N-acetyl-galactosamine-beta-1,3-galactosyltransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8705 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O96024 | |
| B3galt4 | |
| Recombinant protein corresponding to Human B3GALT4. Recombinant protein control fragment (Product #RP-99281). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction