missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B3GALT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA569377
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 93%; Zebrafish: 93%.
This gene is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesized by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). This gene encodes a protein that functions in N-linked glycoprotein glycosylation and shows strict donor substrate specificity for UDP-galactose.
Especificaciones
| B3GALT2 | |
| Polyclonal | |
| Unconjugated | |
| B3GALT2 | |
| B3galt2; Beta-1,3-galactosyltransferase 2; beta-1,3-GalTase 2; BETA3GALT2; beta-3-galt2; beta3Gal-T2; GLCT2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase-II; UDP-galactose:2-acetamido-2-deoxy-D-glucose 3beta-galactosyltransferase 2; Unknown (protein for MGC:127611); zgc:76904; zgc:76904 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26878 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O54905 | |
| B3GALT2 | |
| synthetic peptide corresponding to a region of mouse B3GALT2 (aa 352-401). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido