missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B3GALT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101455
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total B3GALT1.
This gene is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesized by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). This gene is expressed exclusively in the brain. The encoded protein shows strict donor substrate specificity for UDP-galactose. [provided by RefSeq, Jul 2008].
Specifications
| B3GALT1 | |
| Polyclonal | |
| Unconjugated | |
| B3GALT1 | |
| B3galt1; Beta-1,3-galactosyltransferase 1; beta-1,3-GalTase 1; beta3GalT1; beta3Gal-T1; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 1; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase-I; UDP-galactose:beta-N-acetylglucosamine beta1, 3-galactosyltransferase 1; UDP-galactose:beta-N-acetyl-glucosamine-beta-1,3-galactosyltransferase 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26877, 8708 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O54904, Q9Y5Z6 | |
| B3GALT1 | |
| A synthesized peptide derived from human B3GALT1(Accession Q9Y5Z6), corresponding to amino acid residues S6-R56. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction