missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GALT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69327
This item is not returnable.
View return policy
Description
B3GALT1 Polyclonal specifically detects B3GALT1 in Human samples. It is validated for Western Blot.
Specifications
| B3GALT1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| beta-1,3-galactosyltransferase 1, Beta-1,3-GalTase 1, beta-3-galt1, beta3Gal-T1, Beta3GalT1, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.62, MGC126594, UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 1, UDP-galactose:beta-N-acetyl-glucosamine-beta-1,3-galactosyltransferase 1 | |
| Rabbit | |
| 38 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y5Z6 | |
| B3GALT1 | |
| Synthetic peptides corresponding to B3GALT1(UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 1) The peptide sequence was selected from the N terminal of B3GALT1. Peptide sequence MASKVSCLYVLTVVCWASALWYLSITRPTSSYTGSKPFSHLTVARKNFTF The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 8708 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction