missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B3galnt1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569352
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
B3GALNT1 gene ontology annotations related to this gene include protein glycosylation.
Specifications
| B3galnt1 | |
| Polyclonal | |
| Unconjugated | |
| B3GALNT1 | |
| B3galnt1; B3galt3; b3Gal-T3; b3GT3; Beta-1,3-galactosyltransferase 3; beta-1,3-GalNAc-T1; Beta-1,3-GalTase 3; beta-1,3-N-acetylgalactosaminyltransferase 1; beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group); beta-3-GalNAc-T1; beta3GalT3; beta3Gal-T3; beta-3-Gx-T3; Brainiac 1; brainiac1; galactosylgalactosylglucosylceramide beta-D-acetyl-galactosaminyltransferase; galT3; Gb4Cer; GLCT3; GLOB; Globoside blood group; globoside synthase; globotriaosylceramide 3-beta-N-acetylgalactosaminyltransferase; Mbrn 1; Mbrn1; P; P antigen synthase; P blood group globoside; P1; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 1; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase III; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 3; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 3 (Globoside blood group); UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 1; UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 1; UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 1 (Globoside blood group); UDP-N-acetylgalactosamine:globotriaosylceramide beta-1,3-N-acetylgalactosaminyltransferase; UNQ531/PRO1074 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 310508 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6AY39 | |
| B3GALNT1 | |
| synthetic peptide corresponding to a region of rat B3galnt1 (aa 204-253). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction