missing translation for 'onlineSavingsMsg'
Learn More

Brady™ B30 Series Pre-Printed GHS CLP Chemical Labels

Product Code. 30041405
Click to view available options
Dimensions (LxW):
110.00 mm (W) x 75.00 mm (H)
84.99 mm (W) x 55.00 mm (H)
Display Text:
2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker
3 Red Diamonds per Sticker
Unit Size:
Pack of 270
Pack of 340
This item is not returnable. View return policy

Product Code. 30041405

Brand: Brady™ 121643

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pre-printed GHS/CLP chemical labels for BBP3X/S3XXX/i3300 printers.

For BBP3X/S3XXX/i3300 printers.

Specifications

Certifications/Compliance BS 5609:1986 Section 2 and section 3
Description Pre-printed GHS/CLP Chemical Labels for BBP3X/S3XXX/i3300 Printers
Adhesive Type Permanent Acrylic
Coating None
Color Red on White
Finish Gloss
Material Vinyl
Printable Sides Front
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Display Text 2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker
For Use With (Application) Facility Identification, Hazard Communication Identification, Electrical Labelling, Safety Identification
For Use With (Equipment) BBP11, BBP12, PR Plus, THT 25 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
Model B30-R10000
No. per Pack 270 Labels/Roll
Quantity 270 Stickers
Shape Rectangle
Size 110.00 mm (W) x 75.00 mm (H)
Temperature Range (English) -4°F to 167°F
Temperature Range (Metric) -20°C to +75°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (LxW) 110.00 mm (W) x 75.00 mm (H)
Length (English) 2.953 in.
Length (Metric) 75.00 mm
Width (English) 4.331 in.
Width (Metric) 110.00 mm
Width (English) Printable Area 110.00 mm
Alternate Part Number B30-261-7569-CLP4A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.