missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B23/NPM1 Mouse anti-Human, Mouse, Rat, CoraLite 532, Clone: 4F12A3, Proteintech
Shop All Proteintech ProductsDescription
B23/NPM1 Monoclonal antibody apecifically detetects B23/NPM1 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.
Specifications
Specifications
| Antigen | B23/NPM1 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 4F12A3 |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite 532 |
| Formulation | PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. |
| Gene Accession No. | BC002398 |
| Gene Symbols | NPM1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?