missing translation for 'onlineSavingsMsg'
Learn More

B220, MedChemExpress

Product Code. 16490579
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490579 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16490579 Supplier Medchem Express Supplier No. HY100272

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress B220 is an antiviral agent which can inhibit the growth of HSV-1, HSV-2 and human cytomegalovirus (CMV).

B220 is an antiviral agent which can inhibit the growth of HSV-1, HSV-2 and human cytomegalovirus (CMV).

Specifications

Chemical Name or Material B220
Physical Form Solid
CAS 112228-65-6
Color Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C20H22N4
For Use With (Application) Cancer-programmed cell death
SMILES CC1=C(C)C=C2N=C3C(N(CCN(C)C)C4=C3C=CC=C4)=NC2=C1
Molecular Weight (g/mol) 318.42
Formula Weight 318.42
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.