missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B220/CD45R Antibody (HIS24), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43408-0.025MG
This item is not returnable.
View return policy
Description
B220/CD45R Monoclonal specifically detects B220/CD45R in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| B220/CD45R | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| PTPRC | |
| The immunogen for this antibody was CD45R. | |
| 0.025 mg | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 5788 | |
| Store at 4C. Do not freeze. |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
| HIS24 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Rat | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction