missing translation for 'onlineSavingsMsg'
Learn More

B220/CD45R Antibody (F10-89-4), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18428754 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428754 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428754 Supplier Novus Biologicals Supplier No. NBP254455AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

B220/CD45R Monoclonal antibody specifically detects B220/CD45R in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen B220/CD45R
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone F10-89-4
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Host Species Mouse
Immunogen Human T-Lymphocytes (Uniprot: P08575)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.