missing translation for 'onlineSavingsMsg'
Learn More

B16-F10 (mouse skin melanoma) Whole cell lysate, Non-denatured; Abnova

Product Code. 16099635
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16099635 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16099635 Supplier Abnova Supplier No. L060V2.200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Western Blotting, Immunoprecipitation

Specifications

For Use With (Application) Immunoprecipitation, Western Blot
Tissue Melanoma
Description Whole cell lysate (non-denatured)
Lysis Buffer Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
Preparation Method Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay).
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 200 μg
Storage Buffer In modified RIPA Lysis Buffer.
Host Species Mouse
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.