missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B1 Bradykinin Receptor Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%) Gibbon (94%) Rat, Hamster (89%) Mouse, Rabbit, Horse (83%).
Bradykinin, a 9 aa peptide, is generated in pathophysiologic conditions such as inflammation, trauma, burns, shock, and allergy. Two types of G-protein coupled receptors have been found which bind bradykinin and mediate responses to these pathophysiologic conditions. The protein encoded by this gene is one of these receptors and is synthesized de novo following tissue injury. Receptor binding leads to an increase in the cytosolic calcium ion concentration, ultimately resulting in chronic and acute inflammatory responses.
Specifications
Specifications
| Antigen | B1 Bradykinin Receptor |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | Bdkrb1 |
| Gene Accession No. | P46663 |
| Gene Alias | B1 bradykinin receptor; B1BKR; B1R; Bdkrb; BDKRB1; BDKRB2; Bk b1 receptor; BK-1 receptor; BKB1R; BKR; BKR1; BKRB1; BRADYB1; bradykinin B1 receptor; bradykinin receptor 1; bradykinin receptor B1; bradykinin receptor B2; bradykinin receptor, beta 1; KB1; kinin B1; kinin B1 receptor |
| Gene Symbols | Bdkrb1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?