Learn More
BOLA B.safe Direct Filling System
Product Code. 18425782
Change view
Click to view available options
Product Type:
Direct Filling System
Unit Size:
Each
Product Code. Product Type unitSize
18425782 Direct Filling System Each
1 options
Product Code. 18425782 Supplier BOLA Supplier No. R150005
Designed for direct installation into laboratory fume hoods or under-shelf setups for enhanced safety and operational convenience
  • Highly specialized system for the disposal of especially viscous liquids or liquid waste with solid contents
  • b.safe Funnel 130 made of static dissipative PE-EX with GL 45 cap, hinged lid and sieve
  • Safety base cabinet T90 in colour RAL7035 (light gray), outer dimensions: W 593 x D 574 x 600 mm, all passages in fire protection compliant design
  • Telescopic disposal tube made of stainless steel for working height 90 cm with lock nut, canister lid, capacitive fill
  • Level sensor, vent hose and collecting vessel
  • b.safe Isolated Amplifier including power supply
Specifications
Product Type Direct Filling System
Disposable Reusable