missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B-Raf (V600E) Recombinant Rabbit Monoclonal Antibody (10Y9P0)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557885
This item is not returnable.
View return policy
Description
Immunogen sequence: KTRHVNILLF MGYSTKPQLA IVTQWCEGSS LYHHLHIIET KFEMIKLIDI ARQTAQGMDY LHAKSIIHRD LKSNNIFLHE DLTVKIGDFG LATVKSRWSG.
This gene, B-RAF/BRAF, encodes a protein belonging to the RAF family of serine/threonine protein kinases. This protein plays a role in regulating the MAP kinase/ERK signaling pathway, which affects cell division, differentiation, and secretion. Mutations in this gene, most commonly the V600E mutation, are the most frequently identified cancer-causing mutations in melanoma, and have been identified in various other cancers as well, including non-Hodgkin lymphoma, colorectal cancer, thyroid carcinoma, non-small cell lung carcinoma, hairy cell leukemia and adenocarcinoma of lung. Mutations in this gene are also associated with cardiofaciocutaneous, Noonan, and Costello syndromes, which exhibit overlapping phenotypes. A pseudogene of this gene has been identified on the X chromosome.
Specifications
| B-Raf (V600E) | |
| Recombinant Monoclonal | |
| 1.79 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P15056 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 673 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 10Y9P0 | |
| Unconjugated | |
| BRAF | |
| BRAF | |
| A synthetic peptide corresponding to a sequence within amino acids 507-606 of B-Raf (NP_004324.2). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur