Learn More
B-Raf Antibody (CL4004) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59035IR
Product Code. 30292927
  • £558.00 / 0.10mL
Description
B-Raf Monoclonal antibody specifically detects B-Raf in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
B-Raf
Monoclonal
DyLight 755
94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109
This antibody was developed using a recombinant protein derived from P15056, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, Phospho Specific
673
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL4004
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified