Learn More
Europa Bioproducts™ B-Nicotinamide-adenine-dinucleotide Phosphate Tetrasodium Salt
Product Code. 12881680
Encompass
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. 12881680
Brand: Europa Bioproducts™ OR304A0.5g
Used as a coenzyme in dehydrogenase reactions.
Keep tightly closed stoppered in the dark below 5°C. Moisture will accelerate the purity reduction . For prolonged storage keep below -20°C. Solution is acidic and extremely unstable . Most stable at pH 10-11.
Specifications
Quantity 500 mg