missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ B-Myb Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29277
This item is not returnable.
View return policy
Description
B-Myb Polyclonal antibody specifically detects B-Myb in Human samples. It is validated for Immunohistochemistry
Specifications
| B-Myb | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| Unconjugated | |
| B-MYB, BMYBMGC15600, Myb-like protein 2, myb-related protein B, v-myb avian myeloblastosis viral oncogene homolog-like 2, v-myb myeloblastosis viral oncogene homolog (avian)-like 2 | |
| Between 450 and 500 | |
| 25 μg | |
| Apoptosis, Cancer, Core ESC Like Genes, Phospho Specific, Stem Cell Markers | |
| 4605 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction