missing translation for 'onlineSavingsMsg'
Learn More

b-Morpholinopropionitrile, TRC

Product Code. 30466760
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30466760

Brand: Toronto Research Chemicals TRCM7238505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material beta-Morpholinopropionitrile
CAS 4542-47-6
Molecular Formula C7H12N2O
Quantity 5 g
Synonym 3-Morpholinopropanenitrile, 3-Morpholinopropionitrile, N-(2-Cyanoethyl)morpholine, NSC 20547, NSC 41718, 4-Morpholinepropanenitrile
InChI Formula InChI=1S/C7H12N2O/c8-2-1-3-9-4-6-10-7-5-9/h1,3-7H2
SMILES N#CCCN1CCOCC1
IUPAC Name 3-morpholin-4-ylpropanenitrile
Molecular Weight (g/mol) 140.18
Formula Weight 140.095
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.