missing translation for 'onlineSavingsMsg'
Learn More
Learn More
b-Morpholinopropionitrile, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | beta-Morpholinopropionitrile |
| CAS | 4542-47-6 |
| Molecular Formula | C7H12N2O |
| Quantity | 25 g |
| Synonym | 3-Morpholinopropanenitrile, 3-Morpholinopropionitrile, N-(2-Cyanoethyl)morpholine, NSC 20547, NSC 41718, 4-Morpholinepropanenitrile |
| InChI Formula | InChI=1S/C7H12N2O/c8-2-1-3-9-4-6-10-7-5-9/h1,3-7H2 |
| SMILES | N#CCCN1CCOCC1 |
| IUPAC Name | 3-morpholin-4-ylpropanenitrile |
| Molecular Weight (g/mol) | 140.18 |
| Formula Weight | 140.095 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction