missing translation for 'onlineSavingsMsg'
Learn More
Learn More
b-Cyclodextrin Sulfobutyl Ethers, Sodium Salts, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Betadex Sulfobutyl Ether Sodium |
| CAS | 182410-00-0 |
| Molecular Formula | C42H70O35•xNa•x(C4H9O3S) |
| Quantity | 10 g |
| Synonym | Betadex Sulfobutyl Ether Sodium Salt |
| InChI Formula | InChI=1S/C42H70O35.C4H10O3S.Na/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51;1-2-3-4-5-6-7-8;/h8-63H,1-7H2;8H,2-4H2,1H3;/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18?,19?,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40+,41-,42-;;/m1../s1 |
| SMILES | O[C@@H]([C@@H]1O[C@H]([C@@H](C([C@@H]2O[C@H]([C@@H]([C@H]([C@@H]3O[C@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O5)CO)O)O)O[C@@H]3CO)O)O)O[C@@H]2CO)[C@H]([C@H](O[C@@H]1CO)O[C@H]6[C@@H]([C@H]([C@H](O[C@@H]6CO)O[C@H]7[C@@H]([C@H]([C@H](O[C@@H]7CO)O[C@H]8C([C@H]([C@H]5O[C@@H]8CO)O)O)O)O)O)O)O |
| IUPAC Name | β-Cyclodextrin, sulfobutyl ethers, sodium salts |
| Molecular Weight (g/mol) | 1451.3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction