missing translation for 'onlineSavingsMsg'
Learn More

b-AP15, MedChemExpress

Product Code. 16493396
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
25mg
500mg
50mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493396 25 mg 25mg
16492097 50 mg 50mg
16431116 200 mg 200mg
16426386 100 mg 100mg
16421716 500 mg 500mg
16405958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16493396 Supplier Medchem Express Supplier No. HY13989
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress b-AP15 is a specific inhibitor of the deubiquitinating enzymes UCHL5 and Usp14.

b-AP15 is a specific inhibitor of the deubiquitinating enzymes UCHL5 and Usp14.

Specifications

Chemical Name or Material b-AP15
CAS 1009817-63-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H17N3O6
Quantity 25 mg
Synonym NSC 687852
Solubility Information DMSO : 20 mg/mL (47.69 mM; Need ultrasonic)
SMILES O=C1/C(CN(C(C=C)=O)C/C1=C\C2=CC=C([N+]([O-])=O)C=C2)=C/C3=CC=C([N+]([O-])=O)C=C3
Molecular Weight (g/mol) 419.39
Formula Weight 419.39
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.