missing translation for 'onlineSavingsMsg'
Learn More

b-Anilinoethanol, TRC

Product Code. 30348471
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 30348471

Brand: Toronto Research Chemicals TRCA663770100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material beta-Anilinoethanol
CAS 122-98-5
Molecular Formula C8H11NO
Quantity 100 g
Synonym 2-(Phenylamino)ethanol, 2-Anilinoethanol, N-(2-Hydroxyethyl)benzenamine, N-(2-Hydroxyethyl)aniline, N-(2-Hydroxyethyl)phenylamine, N-(beta-Hydroxyethyl)anilineN-Anilinoethanol, N-Ethanolaniline, N-Phenyl-2-aminoethanol, N-Phenylethanolamine, NSC 8393
InChI Formula InChI=1S/C8H11NO/c10-7-6-9-8-4-2-1-3-5-8/h1-5,9-10H,6-7H2
SMILES OCCNc1ccccc1
IUPAC Name 2-anilinoethanol
Molecular Weight (g/mol) 137.18
Formula Weight 137.0841
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.