missing translation for 'onlineSavingsMsg'
Learn More

b-Alanine Tert-butyl Ester Hydrochloride, TRC

Product Code. 30320991
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320991 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320991 Supplier Toronto Research Chemicals Supplier No. TRCA5105655G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material beta-Alanine Tert-butyl Ester Hydrochloride
CAS 58620-93-2
Molecular Formula C7H15NO2 . HCl
Quantity 5 g
Synonym Tert-butyl beta-Alaninate Hydrochloride, Tert-butyl 3-Aminopropanoate Hydrochloride, 3-Aminopropionic Acid Tert-butyl Ester Hydrochloride, 3-Aminopropanoic Acid Tert-butyl Ester Hydrochloride, Beta-alanine Tert-butyl Ester Hydrochloride, beta-Alanine t-Butyl Ester Hydrochloride, beta-Alanine 1,1-Dimethylethyl Ester Hydrochloride
InChI Formula InChI=1S/C7H15NO2.ClH/c1-7(2,3)10-6(9)4-5-8;/h4-5,8H2,1-3H3;1H
SMILES Cl.CC(C)(C)OC(=O)CCN
IUPAC Name tert-butyl 3-aminopropanoate;hydrochloride
Molecular Weight (g/mol) 181.66
Formula Weight 181.087
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.