missing translation for 'onlineSavingsMsg'
Learn More

Brady™ B-7610 Paper Thermal Transfer Labels, 76 mm Core

Product Code. 11598774
Change view
Click to view available options
For Use With (Equipment):
BBP11, BBP12, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
BBP11, BBP12, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Height (Metric):
50 mm
55.9 mm
80 mm
159 mm
Width (Metric):
80 mm
104 mm
108 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Height (Metric) Width (Metric) unitSize
11598774 BBP11, BBP12, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 159 mm 108 mm Each
11569883 BBP11, BBP12, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 50 mm 80 mm Each
11568774 BBP11, BBP12, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 55.9 mm 104 mm Each
11518784 BBP11, BBP12, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 80 mm 108 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11598774 Supplier Brady™ Supplier No. 800865
  • ONLINE EXCLUSIVE PRICE
    £165.00
    £161.95 / Pack of 1
    Save £3.05
    1% Off
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thermal transfer printable paper for low cost general labeling applications.

  • 76 mm core
  • Permanent acrylic adhesive
  • Matte finish

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Paper (B-7610)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification
For Use With (Equipment) BBP11, BBP12, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
Model THT-108x159-7610
No. per Pack 3000 Label(s) / Box
Quantity 3000 Labels
Shape Rectangle
Size 108.00 mm (W) x 159.00 mm (H)
Temperature Range (English) -40°F to 122°F
Temperature Range (Metric) -40°C to +50°C
Thickness (English) 0.0032 in.
Thickness (Metric) 0.081 mm
Dimensions (L x W) 108.00 mm (W) x 159.00 mm (H)
Length (English) 6.26 in.
Width (English) 4.25 in.
Width (Metric) 108 mm
Width (English) Printable Area 108.00 mm
Height (English) 6.26 in.
Height (Metric) 159 mm
Alternate Part Number THT-108x159-7610
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.