missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azurocidin/CAP37/HBP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | Azurocidin/CAP37/HBP |
|---|---|
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699145
|
Novus Biologicals
NBP2-99431-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638055
|
Novus Biologicals
NBP2-99431-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Azurocidin/CAP37/HBP Polyclonal antibody specifically detects Azurocidin/CAP37/HBP in Human samples. It is validated for ELISASpecifications
| Azurocidin/CAP37/HBP | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| 0.2 um filtered solution in PBS | |
| 566 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AZAMP, AZU, azurocidin, azurocidin 1, CAP37Heparin-binding protein, cationic antimicrobial protein 37, HBPCationic antimicrobial protein CAP37, HUMAZUR, NAZC, neutrophil azurocidin | |
| Produced in rabbits immunized with purified, recombinant Human Azurocidin/CAP37/HBP (Accession#: NP_001691.1; Met 1-Pro 250). AZU1 specific IgG was purified by Human AZU1 affinity chromatography . | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title