missing translation for 'onlineSavingsMsg'
Learn More

Azurocidin/CAP37/HBP Antibody, Novus Biologicals™

Product Code. 18336757 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336757 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336757 Supplier Novus Biologicals Supplier No. H00000566B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Azurocidin/CAP37/HBP Polyclonal antibody specifically detects Azurocidin/CAP37/HBP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Azurocidin/CAP37/HBP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_001691.1
Gene Alias AZAMP, AZU, azurocidin, azurocidin 1, CAP37Heparin-binding protein, cationic antimicrobial protein 37, HBPCationic antimicrobial protein CAP37, HUMAZUR, NAZC, neutrophil azurocidin
Host Species Mouse
Immunogen AZU1 (NP_001691.1, 1 a.a. - 251 a.a.) full-length human protein. MTRLTVLALLAGLLASSRAGSSPLLDIVGGRKARPRQFPFLASIQNQGRHFCGGALIHARFVMTAASCFQSQNPGVSTVVLGAYDLRRRERQSRQTFSISSMSENGYDPQQNLNDLMLLQLDREANLTSSVTILPLPLQNATVEAGTRCQVAGWGSQRSGGRLSRFPRFVNVTVTPEDQCRPNNVCTGVLTRRGGICNGDGGTPLVCEGLAHGVASFSLGPCGRGPDFFTRVALFRDWIDGVLNNPGPGPA
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 566
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.