missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Azurocidin/CAP37/HBP Polyclonal specifically detects Azurocidin/CAP37/HBP in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Azurocidin/CAP37/HBP |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | AZAMP, AZU, azurocidin, azurocidin 1, CAP37Heparin-binding protein, cationic antimicrobial protein 37, HBPCationic antimicrobial protein CAP37, HUMAZUR, NAZC, neutrophil azurocidin |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Azurocidin/CAP37/HBP (NP_001691.1; Met 1-Pro 250). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?