missing translation for 'onlineSavingsMsg'
Learn More

Azulene, 99%

Product Code. 11358075
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358075 250 mg 250mg
11348075 1 g 1g
15478879 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358075 Supplier Thermo Scientific Alfa Aesar Supplier No. L08271.MD
  • ONLINE EXCLUSIVE PRICE
    £47.30
    £42.65 / 250mg
    Save £4.65
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 275-51-4 | C10H8 | 128.174 g/mol

Azulene is used as an additive in ointments. It is an active ingredient in cosmetics, shaving creams and other topicals. It is used as antioxidant and in anti-inflammatory activities. It is involved in the colorimetric determination of furfural. It acts as ligand for low valent metals and forms pi complexes with both cyclopentadienyl and cylcloheptatrienyl ligands in organometallic chemistry. It is also used as precursor for the preparation of (azulene)Mo2 (CO)6 and (azulene)Fe2 (CO)5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azulene is used as an additive in ointments. It is an active ingredient in cosmetics, shaving creams and other topicals. It is used as antioxidant and in anti-inflammatory activities. It is involved in the colorimetric determination of furfural. It acts as ligand for low valent metals and forms pi complexes with both cyclopentadienyl and cylcloheptatrienyl ligands in organometallic chemistry. It is also used as precursor for the preparation of (azulene)Mo2 (CO)6 and (azulene)Fe2 (CO)5.

Solubility
Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 275-51-4
Molecular Formula C10H8
Molecular Weight (g/mol) 128.174
MDL Number MFCD00003810
InChI Key CUFNKYGDVFVPHO-UHFFFAOYSA-N
Synonym cyclopentacycloheptene, azunamic, bicyclo 5.3.0 decapentaene, azulen, unii-82r6m9mglp, bicyclo 5.3.0-1,3,5,7,9-decapentaene, bicyclo 5.3.0-deca-2,4,6,8,10-pentaene, 82r6m9mglp, azulekeep
PubChem CID 9231
ChEBI CHEBI:31249
IUPAC Name azulene
SMILES C1=CC=C2C=CC=C2C=C1

Specifications

Melting Point 98°C to 101°C
Boiling Point 241°C to 243°C
Quantity 250 mg
Beilstein 969517
Merck Index 14,926
Solubility Information Soluble in organic solvents.
Formula Weight 128.17
Percent Purity 99%
Chemical Name or Material Azulene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.