missing translation for 'onlineSavingsMsg'
Learn More

Aztreonam, MedChemExpress

Product Code. 16498116
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498116 100 mg 100mg
16452036 500 mg 500mg
16431386 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498116 Supplier Medchem Express Supplier No. HYB0129
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aztreonam (SQ-26,776) is a synthetic monocyclic beta-lactam antibiotic, which has a very high affinity for penicillin-binding protein 3 (PBP-3).

Aztreonam (SQ-26,776) is a synthetic monocyclic beta-lactam antibiotic, which has a very high affinity for penicillin-binding protein 3 (PBP-3).

Specifications

Chemical Name or Material Aztreonam
Physical Form Solid
CAS 78110-38-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H17N5O8S2
For Use With (Application) COVID-19-immunoregulation
Synonym SQ-26,776
Solubility Information DMSO : 50 mg/mL (114.83 mM; Need ultrasonic) ∣H2O : 10 mg/mL (22.97 mM; Need ultrasonic)
SMILES CC(C)(O/N=C(C1=CSC(N)=N1)\C(N[C@H]2[C@H](C)N(S(=O)(O)=O)C2=O)=O)C(O)=O
Molecular Weight (g/mol) 435.43
Formula Weight 435.43
Percent Purity 98.37%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.