missing translation for 'onlineSavingsMsg'
Learn More

Aztreonam Ethyl Ester, TRC

Product Code. 30330939
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330939 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330939 Supplier Toronto Research Chemicals Supplier No. TRCA96521525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aztreonam Ethyl Ester
CAS 102579-57-7
Molecular Formula C15 H21 N5 O8 S2
Quantity 25 mg
Synonym Aztreonam Ethyl Ester
InChI Formula InChI=1S/C15H21N5O8S2/c1-5-27-13(23)15(3,4)28-19-10(8-6-29-14(16)17-8)11(21)18-9-7(2)20(12(9)22)30(24,25)26/h6-7,9H,5H2,1-4H3,(H2,16,17)(H,18,21)(H,24,25,26)/b19-10-/t7-,9-/m0/s1
SMILES CCOC(=O)C(C)(C)O\N=C(/C(=O)N[C@H]1[C@H](C)N(C1=O)S(=O)(=O)O)\c2csc(N)n2
IUPAC Name (2S,3S)-3-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(1-ethoxy-2-methyl-1-oxopropan-2-yl)oxyiminoacetyl]amino]-2-methyl-4-oxoazetidine-1-sulfonic acid
Molecular Weight (g/mol) 463.486
Formula Weight 463.083
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.