missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Cargille™ Immersion Oil

Product Code. 11358113
Change view
Click to view available options
Type:
Immersion Oil Type A
Immersion Oil Type B
Immersion Oil Type FF
Viscosity:
1250 cSt
150 cSt
170 cSt
Quantity:
30 mL
120 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Viscosity Quantity unitSize
11358113 Immersion Oil Type FF 170 cSt 120 mL Each
12423170 Immersion Oil Type B 1250 cSt 30 mL Each
11338113 Immersion Oil Type B 1250 cSt 120 mL Each
11328113 Immersion Oil Type A 150 cSt 120 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11358113 Supplier Azpack™ Supplier No. 11358113

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Choose between three types for normal or fluorescence microscopy. Azpack™ Cargille™ Immersion Oil is supplied with a listing of optical and physical properties.

  • Supplied with specific optical and physical properties listing
  • For use with normal light or fluorescence microscopy

Specifications

Type Immersion Oil Type FF
Viscosity 170 cSt
For Use With (Application) Normal light or fluorescence microscopy
Quantity 120 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.