missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress AZP-531

Product Code. 16432699
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16432699 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16432699 Supplier MedChemExpress Supplier No. HYP0231
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AZP-531 is an analogue of unacylated ghrelin designed to improve glycaemic control and reduce weight.

Specifications

Purity 98.76%
CAS 1088543-62-7
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Metabolism-sugar/lipid metabolism
Molecular Weight (g/mol) 962.02 g/mol
Product Type Peptides
Quantity 1 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (103.95 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.