missing translation for 'onlineSavingsMsg'
Learn More

Azoxystrobin, TRC

Product Code. 30133906
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133906 500 mg 500mg
30040970 1 g 1g
30003920 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133906 Supplier Toronto Research Chemicals Supplier No. TRCA965150500MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131860-33-8
Molecular Formula C22 H17 N3 O5
Molecular Weight (g/mol) 403.39
Synonym Benzeneacetic acid, 2-[[6-(2-cyanophenoxy)-4-pyrimidinyl]oxy]-α-(methoxymethylene)-, methyl ester, (E)-, A 14115, Abound, Abound 2.08F, Amistar, Amistar 250SC, Azaka, Azoxystrobin, Azoxystrobine, Bankit, Cruiser Extreme, Dynasty, Heritage, Heritage 50DF, ICI-A 5504, Methyl (E)-2-[2-[6-(2-cyanophenoxy)pyrimidin-4-yloxy]phenyl]-3-methoxypropenoate, Methyl (E)-2-[2-[[6-(2-cyanophenoxy)pyrimidin-4-yl]oxy]phenyl]-3-methoxyacrylate, Onestar, Ortiva, Priori, Protege, Protege FL, Quadris
IUPAC Name methyl (E)-2-[2-[6-(2-cyanophenoxy)pyrimidin-4-yl]oxyphenyl]-3-methoxyprop-2-enoate
SMILES CO\C=C(\C(=O)OC)/c1ccccc1Oc2cc(Oc3ccccc3C#N)ncn2

Specifications

Analyte or Component Names Azoxystrobin
Shipping Condition Room Temperature
InChI Formula InChI=1S/C22H17N3O5/c1-27-13-17(22(26)28-2)16-8-4-6-10-19(16)30-21-11-20(24-14-25-21)29-18-9-5-3-7-15(18)12-23/h3-11,13-14H,1-2H3/b17-13+
Solution Type Neat
Industry Type Pharmaceutical
Quantity 500 mg
Formula Weight 403.1168
Percent Purity >95%
Chemical Name or Material Azoxystrobin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.